Submersible Aquarium Heater Thermostat
Supplier: ROLF C. HAGEN (USA) CORP.
A782 A784 A783
470021-438EA 44 USD
470021-438 470021-440 470014-986
Submersible Aquarium Heater Thermostat
Specimens Live Specimens Aquaria - Living Material
Waterproof Unit Is Completely Safe, Even When Wet
Install this heater/thermostat anywhere in your tank; suction cups will hold the instrument securely against the glass. It features a temperature-set dial, ceramic heater core, and pilot light. It will work in both freshwater and saltwater aquariums. Temperature range: 64–88°F. 115 VAC. Size: 1" dia. When ordering, allow 5 W per gallon of water, 10 W per gallon if room temperature fluctuates greatly.