PYREX® Heavy-Duty-Grade Beakers
Supplier: Corning
1003-150 1003-1L 1003-250 1003-2L 1003-400 1003-4L 1003-600
470211-336CS 637.8 USD
470211-336 470211-338 470211-340 470211-342 470211-344 470211-346 470211-348
PYREX® Heavy-Duty-Grade Beakers
Beakers
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Good temperature resistance
  • With extra large marking spot
Heavy-duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanised washing operations. Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content.
Certificaciones: Meet ASTM E960 specifications.