BioMixer™ Nutating Rockers
Supplier: Ward's
B3D1020_WARD B3D1320_WARD
470133-480EA 530.5 USD
470133-480 470133-482
BioMixer™ Nutating Rockers
Shakers and Mixers Rotating/Rocking Shakers
Economical and Durable 3-Dimensional Mixing
  • Tilt angle up to 20 degrees
  • Cold room and incubator safe
  • 24% more work surface than traditional nutating mixers
  • Capable of loads up to 2 kg
Información para pedidos: Two year warranty.