Glow Chemistry Kit
Supplier: REBECCAS
GP16BRITEG GP16BRITEB GP16BRITER
470106-824EA 4.4 USD
470106-824 470106-826 470106-828
Glow Chemistry Kit
Educational Classroom Kits and Activities
Discover the Chemistry Behind Luminescence
  • Easy to use
  • For grades 11-12
  • Replacement light sticks available separately, in various colors
This demonstration lets you mix sodium hydroxide, dimethyl sulphoxide and luminol to produce a strong blue-green light. Even after the light fades, shaking the bottle will cause more of the luminol to react. Use the supplied light stick to teach students about practical applications of luminescence. Developed by Lynn Hesse.
Información para pedidos: Light sticks have a shelf life of 18 months.
Información de suministro: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.