Alligator Clip/Banana Plug Cords
Supplier: United Scientific Supplies
WAB024-R WAB024-B
470189-210EA 6.3 USD
470189-210 470189-212
Alligator Clip/Banana Plug Cords
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Easily connect a banana plug to a bare connector.
  • Flexible plastic coated wire
  • 15 Amp rating
Cords, 61 cm long, with an alligator clip at one end and a two way banana plug at the other. The alligator clip has a protective vinyl plastic sheath. The flexible plastic coated wire has a rating of 15 A, AC.
Order Now
SPECIFICATIONS
Environmentally Preferable