'Photon' Solar Racer Kit
Supplier: SOLAR MADE SE
PHOTON
470110-950EA 51 USD
470110-950
'Photon' Solar Racer Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Comprehensive and Easy to Assemble
  • Powered by a solar array
  • Maximum velocity of 0.5m/s on a flat surface
  • Customizable plastic frame
Have your students build their own Solar Racer! This easy-to-assemble kit comes with everything needed, including high-powered solar cell and a 1/2 amp solar array and high amp motor for speedy acceleration. Transparent plastic body can be custom painted. When exposed to full sun, the photon racer travels 3m in 6 secs on a smooth, flat surface. Size: 21.5cm L x 10.2cm W x 5.1cm H.