DuPont™ Tychem® 5000 Encapsulated Level B Suits with Airline Access, Rear Entry
Supplier: DuPont
C3526TTNMD000600 C3526TTNXL000600 C3526TTN3X000600 C3526TTN4X000600 C3526TTNLG000600 C3526TTNSM000600 C3526TTN2X000600
89207-842CS 1234.09 USD
89207-842 89207-846 89207-836 89207-838 89207-840 89207-844 89207-834
DuPont™ Tychem® 5000 Encapsulated Level B Suits with Airline Access, Rear Entry
Disposable Garments
Tychem® CPF 3 fabric is a multi-layer film barrier laminated to a durable polypropylene substrate.
  • Flat back
  • Rear entry
  • Standard, single-layered 20 mil PVC visor
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Airline access
  • One exhaust vent
  • Taped seams
This strong and durable material is ideally suited for rigorous activities and rugged situations involving liquid splash or chemical handling
The encapsulated suit design provides protection from liquid splash exposure for both the wearer and respiratory equipment. A flat back design allows use with an airline, and the rear entry closure eliminates direct contact with frontal chemical exposure. A standard 20mil PVC faceshield also provides coverage from overhead liquid splash (faceshield does not provide vapor protection). Taped seams ensure reliable chemical resistance to heavy liquid splashes. The sewn seam is covered with a strip of compatible chemical-resistant material through heatsealing. A storm flap covers the zipper, and can be further secured by rugged hook and loop material to prevent intrusion. Additional standard features include elastic wrist openings, attached socks with outer boot flaps, and one exhaust vent.
The CPF 3 fabric provides protection against more than 149 chemical challenges. These fully encapsulated suits are best suited for chemical handling, petro-chemical applications, hazardous material/waste cleanup, fire department use, industrial HazMat applications, and utility and domestic preparedness applications.
Certificaciones: Thickness meets ASTM D1117.