DL-α-Tocopherol acetate USP
Supplier: SPECTRUM CHEMICAL MFG CORP
VI140-20KGBL
VI140-1LTGL
VI140-100ML
700009-416EA
8363.83
USD
700009-416
95036-558
95036-556
DL-α-Tocopherol acetate USP
DL-α-Tocopherol acetate
Vitamin E Acetate, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₃₁H₅₂O₃ Density: 0.96 Storage Temperature: Ambient |
MDL Number:
MFCD00072042 CAS Number: 7695-91-2 EINECS: 231-710-0 |
Specification Test Results
Assay | 96% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...