Triethanolamine hydrochloride ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

TR116-100GM TR116-25KGBL TR116-500GM TR116-2.5KG
700006-952EA 101.75 USD
700006-952 700006-956 700006-958 700006-954
Triethanolamine hydrochloride ≥99%, Reagent Grade
Triethanolamine hydrochloride

Triethanolamine Hydrochloride, Reagent is a strong base and is as a complexing agent for aluminum ions in aqueous solutions. This reaction is often used to mask such ions before complexometric titrations with another chelating agent such as EDTA. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₆H₁₆ClNO₃
MW: 185.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012596
CAS Number: 637-39-8
EINECS: 211-284-2

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR