L-Threonine ≥98.5%, crystals USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

T1056-5GM T1056-100GM T1056-1KG T1056-500GM T1056-5KG T1056-25GM T1056-50KGBL
95034-092EA 84.1 USD
95034-092 95034-084 95034-086 95034-090 700009-220 95034-088 700009-218
L-Threonine ≥98.5%, crystals USP
L-Threonine

Threonine, Crystal, USP is an amino acid that is used in the biosynthesis of proteins. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR