Sodium carbonate monohydrate ≥99.5% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

SO150-2.5KG SO150-500GM SO150-12KG
95035-750EA 517.3 USD
95035-750 95035-752 700001-798
Sodium carbonate monohydrate ≥99.5% NF
Sodium carbonate monohydrate

Sodium Carbonate, Hydrous, NF is a pH regulator excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Información para pedidos: Allergen statement (available upon request)

Formula: Na₂CO₃·1H₂O
MW: 124.00 g/mol
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6
EINECS: 207-838-8

Order Now

Specification Test Results

Assay 99.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR