Silicone fluid, 1000 cSt 1000
Supplier: Spectrum Chemical Mfg. Corp.
Silicone Fluid, 1000 is a polydimethylsiloxane with good lubrication characteristics for plastics and rubber devices and instruments.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
0.97 g/cm³ (20 °C) Flash Pt: 321 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00084411 CAS Number: 63148-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...