Silicone fluid, 100 cSt 100
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dimethylpolysiloxane, PDMS, Dimethyl siloxane polymer (methyl silicones), Silicone oil, Dimethicone, Polydimethylsiloxane
SI110-1KG
SI110-100GM
700015-848EA
380.35
USD
700015-848
700015-846
Silicone fluid, 100 cSt 100
Silicone fluid
Silicone Fluid, 100 is a polydimethylsiloxane with good lubrication characteristics for plastics and rubber devices and instruments.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
0.965 g/cm³ (20 °C) Flash Pt: 321 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00084411 CAS Number: 63148-62-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...