Potassium citrate monohydrate 99.0-100.5%, granular USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

PO175-12KGPL PO175-500GMP PO175-2.5KGP PO175-2.5KGP
700006-200EA 1312.4 USD
700006-200 95035-152 95035-150 700006-202
Potassium citrate monohydrate 99.0-100.5%, granular USP suitable for use as excipient
Potassium citrate monohydrate

Potassium Citrate, Monohydrate, Granular, USP belongs to a class of drugs known as urinary alkalinizers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: HOC(COOK)(CH₂COOK)₂·H₂O
Melting Pt: 230 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 13,07706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR