D-Panthenol ≥98% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Dexpanthenol, FCC is the provitamin of vitamin B5 and is used in nutritional supplements. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₉H₁₉NO₄ MW: 205.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065006 CAS Number: 81-13-0 EINECS: 201-327-3 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...