DL-Panthenol ≥99% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

P1910-25KGBL P1910-100GM P1910-25GM P1910-500GM
700009-346EA 5345.6 USD
700009-346 95032-992 95032-994 95032-996
DL-Panthenol ≥99% USP
DL-Panthenol

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002944
CAS Number: 16485-10-2
EINECS: 240-540-6

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR