Potassium bromate ≥99% FCC

Supplier: Spectrum Chemical Mfg. Corp.
Danger

P1278-2.5KG P1278-500GM P1278-125GM
95030-892EA 997.2 USD
95030-892 95030-896 95030-894
Potassium bromate ≥99% FCC
Potassium bromate

Potassium Bromate, FCC is considered an oxidizing agent and has been used in the production of malt barley. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: KBrO₃
Melting Pt: 350 °C
Density: 3.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR