Nitrilotriacetic acid trisodium salt ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
N2923-1KG
N2923-500GM
700007-376EA
65.2
USD
700007-376
700007-374
700007-378
Nitrilotriacetic acid trisodium salt ≥99%
Nitrilotriacetic acid trisodium salt
Nitrilotriacetic Acid Trisodium Salt, also known as Triglycine, is an aminopolycarboxylic acid used as a chelating agent and for water softening; it can also be used as a replacement to sodium and potassium triphosphate in cleansers and detergents.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₆NNa₃O₆ MW: 257.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064231 CAS Number: 5064-31-3 EINECS: 225-768-6 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...