N-Methylmorpholine-N-oxide ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide

M2721-25KGBL M2721-25GM M2721-5GM M2721-100GM
700009-180EA 11735.9 USD
700009-180 700009-178 700009-182 700009-176
N-Methylmorpholine-N-oxide ≥97%
N-Methylmorpholine-N-oxide

4-Methylmorpholine N-Oxide is an organic compound used as a sacrificial catalyst and co-oxidant in oxidation reactions in organic chemistry.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005947
CAS Number: 7529-22-8
EINECS: 231-391-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR