Erythrosin B disodium salt
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
C.I. 45430
FD135-20KGBL
FD135-100GM
FD135-25GM
FD135-5KGBL
FD135-500GM
700010-052EA
11383.1
USD
700010-052
700010-050
700010-054
700010-058
700010-056
Erythrosin B disodium salt
Erythrosin B disodium salt
FD and C Red No. 3 | Non-graded FD and C Red No. 3 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂₀H₆I₄Na₂O₅ MW: 879.86 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00144257 CAS Number: 16423-68-0 EINECS: 240-474-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...