Disodium fumarate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Disodium Fumarate, Fumaric acid disodium salt, Fumaric Acid Disodium Salt, Sodium fumarate
F2075-100GM
F2075-500GM
700005-916EA
180.45
USD
700005-916
700005-918
700005-920
Disodium fumarate
Disodium fumarate
Fumaric Acid Disodium Salt, similar to fumaric acid, is a white crystalline salt and has been evaluated as a food additive.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₄H₂Na₂O₄ MW: 160.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064567 CAS Number: 17013-01-3 EINECS: 241-087-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...