Erythrosin B disodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: C.I. 45430

ER125-25GM ER125-5GM
700010-040EA 146.05 USD
700010-040 700010-042
Erythrosin B disodium salt
Erythrosin B disodium salt

Erythrosin B, also known as Red No. 3, can be used in food coloring or as a biological stain.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
EINECS: 240-474-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR