Calcium-D-gluconate monohydrate ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate

CA146-500GM CA146-2.5KG CA146-50KGBL CA146-12KG
95032-276EA 175.45 USD
95032-276 95032-274 89078-664 700006-284
Calcium-D-gluconate monohydrate ≥98.5% USP
Calcium-D-gluconate monohydrate

Calcium Gluconate, Monohydrate, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178
Storage Temperature: Ambient
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR