Curcumin

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

C1452-100GM C1452-25GM C1452-10GM
700004-134EA 615.4 USD
700004-134 700004-138 700004-136
Curcumin
Curcumin

Curcumin, also known as Diferuloylmethane, is used for boron quantification in the curcumin method.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR