Creatinine NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

C1403-100GM C1403-25GM C1403-500GM C1403-12KGBL C1403-1KGBL
95032-710EA 285.75 USD
95032-710 95032-714 95032-716 89078-778 95032-712
Creatinine NF
Creatinine

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₄H₇N₃O
MW: 113.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR