Brij® L23

Supplier: Spectrum Chemical Mfg. Corp.
Warning

BR105-12KG BR105-2.5KG BR105-25GM BR105-500GM
700010-922EA 1146.95 USD
700010-922 700010-924 700010-926 700010-928
Brij® L23
Brij® L23

Brij(R) L23 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.

Formula: (C₂₀H₄₂O₅)n
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00043063
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR