Brij® S100
Supplier: Spectrum Chemical Mfg. Corp.
B1593-25GM
B1593-500GM
B1593-2.5KG
700012-410EA
185.9
USD
700012-410
700012-412
700012-406
Brij® S100
Brij® S100
Brij(R) S100, Polyethylene (100) Stearyl Ether, functions as an aqueous dispersant and oil/water emulsifier.
Formula:
C₁₈H₃₇(OCH₂CH₂)nOH,n~100 Storage Temperature: Ambient |
MDL Number:
MFCD00074177 CAS Number: 9005-00-9 EINECS: 500-017-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...