Spherical to Standard Taper Joint Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5020-10 5020-20 5020-25 5020-30 5020-32 5020-22 5020-21 5020-23 5020-31 5020-33 5020-40 5020-41 5020-42 5020-43 5020-44 5020-45 5020-46 5020-47 5020-48 5020-49 5020-50 5020-51 5020-52 5020-53 5020-54 5020-55
89051-034EA 83.88 USD
89051-034 89051-036 89051-038 89051-040 89051-042 89082-556 10767-742 10767-744 10767-746 10767-748 10767-750 10767-752 10767-754 10767-756 10767-758 10767-760 10767-762 10767-872 10767-874 10767-876 10767-878 10767-880 10767-882 10767-884 10767-886 10767-888
Spherical to Standard Taper Joint Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These connecting adapters feature a spherical joint and an inner standard taper joint.
  • Top spherical joint
  • Bottom [ST] inner joint
Adapters feature a spherical top and a Standard Taper male bottom joint. Spherical joint is either ball or socket.