Striped Safety Tapes, NMC (National Marker Company)
Supplier: ACCUFORM MANUFACTURING, INC
T20718 T20736 T20818 T20836 T20918 T20936 T22018 T22036 T22118 T22136 T22318 T22336 T22418 T22436 T22818 T22836 T22918 T22936
94022-026EA 34.04 USD
94022-026 94022-028 94022-032 94022-034 94022-038 94022-040 94022-048 94022-050 94022-052 94022-054 94022-056 94022-058 94022-060 94022-062 94022-064 94022-066 94022-068 94022-070
Striped Safety Tapes, NMC (National Marker Company)
Tapes
Ideal for floor markings in high traffic areas
  • Ideal for floor markings in high-traffic areas
  • Self-adhesive vinyl tapes designed for long-lasting use
  • Apply indoors or outdoors on smooth, clean, dry surfaces
  • Waterproof and wear-resistant 5mil vinyl tape comes in two sizes and various colors
These self-adhesive vinyl tapes are designed for long-lasting use. Apply indoors or outdoors on smooth, clean dry surfaces. Waterproof and wear-resistant 5 mil vinyl tape comes in two sizes and various colors. Apply self-adhesive backing to clean, smooth dry surface.More Product Information