Precoiled Pipe Markers- Blank Green, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A6004 B6004
94003-528EA 17.79 USD
94003-528 94004-302
Precoiled Pipe Markers- Blank Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certificaciones: Meets ANSI A13.1-2007