Precoiled Pipe Markers 'Condensate', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4024 B4024 C4024 D4024 E4024 F4024 G4024 H4024
94003-328EA 18.34 USD
94003-328 94004-102 94005-498 94009-410 94012-590 94013-786 94014-332 94015-112
Precoiled Pipe Markers 'Condensate', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed an reapplied in other areas.
Certificaciones: Meets ANSI A13.1-2007