Triisopropylsilane

Supplier: CHEM-IMPEX INTERNATIONAL, INC

01966-100G 01966-25G
CH01966-100GEA 80 USD
CH01966-100G CH01966-25G
Triisopropylsilane
Triisopropylsilane
Formula: C₉H₂₂Si
Boiling Pt: 84 °C (1013 hPa)
Density: 0.7734 g/cm³ (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00009657
CAS Number: 6485-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR