'V' Assembly for Rotary Evaporators, Ace Glass Incorporated
Supplier: Ace Glass
3951-05 3951-10
89083-088EA 1563.74 USD
89083-088 89083-092
'V' Assembly for Rotary Evaporators, Ace Glass Incorporated
Condensers
Complete "V" Assembly glassware set for many popular bench-scale rotary evaporators
Fabricated from borosilicate glass. Fits models 200/205 and Series 114-144. Available poly coated for safety, or uncoated. Includes the main glass condenser body with inner coil for vertical installation and heavy wall 1L receiving flask. The bottom condenser joint is spherical [SJ] 35/20 ball joint. The included round-bottom receiving flask has a [SJ] 35/20 socket joint. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.
Información para pedidos: Includes o-ring, 89059-774 GL thread caps, and 89059-810 angled hose connectors. The bottom joint is spherical [SJ] 35/20.