Aluminum Seals, Labconco®
Supplier: Labconco
7577010 7577110
26676-353PK 22.5 USD
26676-353 26676-355
Aluminum Seals, Labconco®
Closures Chromatography Vial Closures
Aluminum Seal
Package of 100. Forms a tight seal around top of Split Stopper and lip of Serum Bottle when crimped. Specifically designed for lyophilization applications. For use with FreeZone Free Dry Systems. Seals are for serum bottles with 20 mm corkage or 13 mm corkage, as indicated.
Order Now
SPECIFICATIONS
Environmentally Preferable