2-Chloro-5-pyridineboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid

L20388-06 L20388-03
AAAL20388-06EA 94.5 USD
AAAL20388-06 AAAL20388-03 AAAL20388-MD
2-Chloro-5-pyridineboronic acid 96%
2-Chloro-5-pyridineboronic acid

Purity: 96% CAS No.: 444120-91-6 Molecular Formula: C5H5BClNO2 Formula Weight: 157.37 Melting Point: 154-157° Beilstein Reference: 9119810

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD03094998
CAS Number: 444120-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR