1,4-Naphthalenedicarboxylic acid 98+%

Warning

Synonyms: Naphthalene-1,4-dicarboxylic acid

L20249-14 L20249-22
AAAL20249-14EA 84.5 USD
AAAL20249-14 AAAL20249-22
1,4-Naphthalenedicarboxylic acid 98+%
1,4-Naphthalenedicarboxylic acid

Purity: 98+% CAS No.: 605-70-9 Molecular Formula: C12H8O4 Formula Weight: 216.19 Melting Point: 321-325° Beilstein Reference: 2051255

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 321…325 °C
Storage Temperature: Ambient
MDL Number: MFCD00014312
CAS Number: 605-70-9
EINECS: 210-094-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR