3-Cyanobenzenesulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride

L20169-03 L20169-06
AAAL20169-03EA 78.7 USD
AAAL20169-03 AAAL20169-06
3-Cyanobenzenesulphonyl chloride 97%
3-Cyanobenzenesulphonyl chloride

Purity: 97% CAS No.: 56542-67-7 Molecular Formula: C7H4ClNO2S Formula Weight: 201.63 Flash Point: 110°(230°F) Melting Point: 47-52° Boiling Point: 114-116°/0.2mm Beilstein Reference: 2832925 UN No.: 3261 MDL No.: MFCD00203480

Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Boiling Pt: 114…116 °C/0.2 mm
Melting Pt: 47…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR