2-Iodo-m-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19876-03
L19876-06
AAAL19876-03EA
52.2
USD
AAAL19876-03
AAAL19876-06
2-Iodo-m-toluic acid 98%
2-Iodo-m-toluic acid
Purity: 98%
CAS No.: 108078-14-4
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Melting Point: 150-153°
Beilstein Reference: 2439521
Formula:
C₈H₇IO₂ Melting Pt: 150…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079764 CAS Number: 108078-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...