Furan-3-boronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Furanboronic acid, 3-Furanylboronic acid
L19834-03
L19834-06
AAAL19834-03EA
88.03
USD
AAAL19834-03
AAAL19834-06
Furan-3-boronic acid 97%
Furan-3-boronic acid
Purity: 97%
CAS No.: 55552-70-0
Molecular Formula: C4H5BO3
Formula Weight: 111.89
Melting Point: 139-144°
Beilstein Reference: 1635653
Formula:
C₄H₅BO₃ MW: 111.89 g/mol Melting Pt: 139…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319007 CAS Number: 55552-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...