2,3-Dimethoxypyridine

Supplier: Thermo Scientific Chemicals
Warning

L19624-06 L19624-14 L19624-03
AAAL19624-06EA 104 USD
AAAL19624-06 AAAL19624-14 AAAL19624-03
2,3-Dimethoxypyridine
2,3-Dimethoxypyridine

CAS No.: 52605-97-7 Molecular Formula: C7H9NO2 Formula Weight: 139.15 Refractive Index: 1.5205 Boiling Point: 98-100°/16mm Beilstein Reference: 1526067

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 98…100 °C/16 mm
Density: 1.104
Storage Temperature: Ambient
MDL Number: MFCD03095043
CAS Number: 52605-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR