4-Methylphenylboronic acid neopentyl glycol ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methylphenylboronic acid neopentyl ester, 5,5-Dimethyl-2-(p-tolyl)-1,3,2-dioxaborinane, 4-Methylbenzeneboronic acid neopentyl glycol ester

L19563-06 L19563-03
AAAL19563-06EA 110 USD
AAAL19563-06 AAAL19563-03
4-Methylphenylboronic acid neopentyl glycol ester 99%
4-Methylphenylboronic acid neopentyl glycol ester

Purity: 99% CAS No.: 380481-66-3 Molecular Formula: C12H17BO2 Formula Weight: 204.08 Melting Point: 93-95° MDL No.: MFCD03788728

Formula: C₁₂H₁₇BO₂
MW: 204.08 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD03788728
CAS Number: 380481-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR