3,5-Difluoropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19539-06
L19539-03
AAAL19539-06EA
162
USD
AAAL19539-06
AAAL19539-03
3,5-Difluoropyridine 98%
3,5-Difluoropyridine
Purity: 98%
CAS No.: 71902-33-5
Molecular Formula: C5H3F2N
Formula Weight: 115.08
Flash Point: 9°(48°F)
Refractive Index: 1.4437
Boiling Point: 92-93°
Beilstein Reference: 7914363
UN No.: 1993
MDL No.: MFCD03412222
Formula:
C₅H₃F₂N MW: 115.08 g/mol Boiling Pt: 92…93 °C Density: 1.268 g/cm³ (20 °C) Flash Pt: 9 °C (48 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03412222 CAS Number: 71902-33-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...