2-Fluoronicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Cyano-2-fluoropyridine
L19538-03
L19538-06
AAAL19538-03EA
32.9
USD
AAAL19538-03
AAAL19538-06
2-Fluoronicotinonitrile 98%
2-Fluoronicotinonitrile
Purity: 98%
CAS No.: 3939-13-7
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Melting Point: 27-30°
Beilstein Reference: 386567
Formula:
C₆H₃FN₂ MW: 122.1 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: 27…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095082 CAS Number: 3939-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...