2-Quinolinecarbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-2-carbonitrile

L19427-06 L19427-03
AAAL19427-06EA 134 USD
AAAL19427-06 AAAL19427-03
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile

Purity: 97% CAS No.: 1436-43-7 Molecular Formula: C10H6N2 Formula Weight: 154.17 Odor: Odorless Melting Point: 93-96° Boiling Point: 160°/23mm Beilstein Reference: 115213 MDL No.: MFCD00134341

Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Boiling Pt: 160 °C/23 mm
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00134341
CAS Number: 1436-43-7
EINECS: 215-865-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR