4-Bromoindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1H-indole

L19417-03 L19417-06
AAAL19417-MDEA 31.76 USD
AAAL19417-MD AAAL19417-03 AAAL19417-06
4-Bromoindole 98%
4-Bromoindole

Purity: 98% CAS No.: 52488-36-5 Molecular Formula: C8H6BrN Formula Weight: 196.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6545 Melting Point: 17° Boiling Point: 88-90°/0.1mm Beilstein Reference: 114891 MDL No.: MFCD00671502

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 88…90 °C/0.1 mm
Melting Pt: 17 °C
Density: 1.563
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671502
CAS Number: 52488-36-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR