4-Iodobenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Iodophenyl)methanol
L19278-06
L19278-03
AAAL19278-06EA
123
USD
AAAL19278-06
AAAL19278-03
4-Iodobenzyl alcohol 97%
4-Iodobenzyl alcohol
Purity: 97%
CAS No.: 18282-51-4
Molecular Formula: C7H7IO
Formula Weight: 234.04
Melting Point: 72-74°
Beilstein Reference: 1931621
Formula:
C₇H₇IO Melting Pt: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD01732720 CAS Number: 18282-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...