6-Methoxyindole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
L19074-03
L19074-MD
AAAL19074-03EA
208
USD
AAAL19074-03
AAAL19074-MD
6-Methoxyindole 98+%
6-Methoxyindole
Purity: 98+%
CAS No.: 3189-13-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 90-94°
Boiling Point: 105°/0.2mm
MDL No.: MFCD00022780
Formula:
C₉H₉NO MW: 147.18 g/mol Boiling Pt: 105 °C/0.2 mm Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022780 CAS Number: 3189-13-7 EINECS: 221-689-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...