2-Methyl-m-anisic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methoxy-2-methylbenzoic acid
L18729-09
L18729-18
AAAL18729-09EA
25.4
USD
AAAL18729-09
AAAL18729-18
2-Methyl-m-anisic acid 97%
2-Methyl-m-anisic acid
Purity: 97%
CAS No.: 55289-06-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
Beilstein Reference: 510395
MDL No.: MFCD02094039
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02094039 CAS Number: 55289-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...