(S)-(+)-3-Chloro-1,2-propanediol 98%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(2S)-(+)-3-Chloropropane-1,2-diol, (S)-(+)-3-Chloro-1,2-dihydroxy propane
L18556-06
L18556-14
AAAL18556-06EA
72.4
USD
AAAL18556-06
AAAL18556-14
AAAL18556-03
(S)-(+)-3-Chloro-1,2-propanediol 98%, ee 98%
(S)-(+)-3-Chloro-1,2-propanediol
Purity: 98%
CAS No.: 60827-45-4
Molecular Formula: C3H7ClO2
Formula Weight: 110.54
Flash Point: 126°(258°F)
Refractive Index: 1.4800
Boiling Point: 213°
Beilstein Reference: 2202929
Optical Rotation: +0.9°(neat)
UN No.: 2689
MDL No.: MFCD00210270
Formula:
ClCH₂CHOHCH₂OH MW: 110.54 g/mol Boiling Pt: 213 °C Density: 1.322 g/cm³ (20 °C) Flash Pt: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210270 CAS Number: 60827-45-4 UN: 2689 ADR: 6.1,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...