3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18433-06
L18433-03
AAAL18433-06EA
282
USD
AAAL18433-06
AAAL18433-03
3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade
3-(Dimethylamino)benzoyl chloride hydrochloride
Purity: 90%
CAS No.: 117500-61-5
Molecular Formula: C9H10ClNO.HCl
Formula Weight: 220.10
Melting Point: 131-133°
Beilstein Reference: 2803337
UN No.: 3261
MDL No.: MFCD02094023
Formula:
C₉H₁₁Cl₂NO MW: 220.1 g/mol Melting Pt: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD19442144 CAS Number: 117500-61-5 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...